3-(5-Bromo-2-methyl-4-nitro-1H-imidazol-1-yl)propanenitrile

Lieferant: Apollo Scientific

OR21879-10G OR21879-1G OR21879-5G
APOSOR21879-10GEA 346 EUR
APOSOR21879-10G APOSOR21879-1G APOSOR21879-5G
3-(5-Bromo-2-methyl-4-nitro-1H-imidazol-1-yl)propanenitrile
3-(5-Bromo-2-methyl-4-nitro-1H-imidazol-1-yl)propanenitrile
Formel: C₇H₇BrN₄O₂
Molecular Weight: 259,06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173665
CAS-Nummer: 139975-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR