5-Methoxyquinazoline-2,4-diamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4-Diamino-5-methoxyquinazoline, 2,4-Diamino-5-methoxy-1,3-benzodiazine
OR2192-500MG
APOSOR2192-500MGEA
184
EUR
APOSOR2192-500MG
5-Methoxyquinazoline-2,4-diamine
5-Methoxyquinazoline-2,4-diamine
Formel:
C₉H₁₀N₄O Molecular Weight: 190,202 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00665830 CAS-Nummer: 27018-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...