2-Benzyl-4,6-dichloropyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[(4,6-Dichloropyrimidin-2-yl)methyl]benzene, 2-Benzyl-4,6-dichloro-1,3-diazine
OR22106-1G
OR22106-5G
APOSOR22106-1GEA
458
EUR
APOSOR22106-1G
APOSOR22106-5G
2-Benzyl-4,6-dichloropyrimidine
2-Benzyl-4,6-dichloropyrimidine
Formel:
C₁₁H₈Cl₂N₂ Molecular Weight: 239,101 g/mol Schmelzpunkt: 56…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00102149 CAS-Nummer: 3740-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...