3-Amino-2-benzylpropanoic acid, N-BOC protected 98%

Lieferant: Apollo Scientific
Achtung

OR2225-1G OR2225-250MG OR2225-5G
APOSOR2225-1GEA 0 EUR
APOSOR2225-1G APOSOR2225-250MG APOSOR2225-5G
3-Amino-2-benzylpropanoic acid, N-BOC protected 98%
3-Amino-2-benzylpropanoic acid, N-BOC protected
Formel: C₁₅H₂₁NO₄
Schmelzpunkt: 94…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03003690
CAS-Nummer: 26250-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR