3,6-Dichloro-9-(oxiran-2-ylmethyl)-9H-carbazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3,6-Dichloro-9H-carbazol-9-yl)-1,2-propenoxide

OR22291-1G OR22291-250MG
APOSOR22291-1GEA 481 EUR
APOSOR22291-1G APOSOR22291-250MG
3,6-Dichloro-9-(oxiran-2-ylmethyl)-9H-carbazole 95%
3,6-Dichloro-9-(oxiran-2-ylmethyl)-9H-carbazole
Formel: C₁₅H₁₁Cl₂NO
Molecular Weight: 292.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00218276
CAS-Nummer: 65679-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR