3-Amino-6-methoxy-1-methyl-1H-indazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methoxy-1-methyl-1H-indazol-3-amine

OR2266-1G OR2266-100MG OR2266-250MG
APOSOR2266-1GEA 560 EUR
APOSOR2266-1G APOSOR2266-100MG APOSOR2266-250MG
3-Amino-6-methoxy-1-methyl-1H-indazole
3-Amino-6-methoxy-1-methyl-1H-indazole
Formel: C₉H₁₁N₃O
Molecular Weight: 177,203 g/mol
Schmelzpunkt: 164…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09998750
CAS-Nummer: 1031927-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR