3-(1H-Imidazol-1-yl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-Formylphenyl)-1H-imidazole, 3-(1-Imidazolyl)benzaldehyde

OR2268-1G OR2268-25G OR2268-5G
APOSOR2268-1GEA 183 EUR
APOSOR2268-1G APOSOR2268-25G APOSOR2268-5G
3-(1H-Imidazol-1-yl)benzaldehyde
3-(1H-Imidazol-1-yl)benzaldehyde
Formel: C₁₀H₈N₂O
Molecular Weight: 172.183 g/mol
Schmelzpunkt: 80…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09836178
CAS-Nummer: 127404-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR