Bicyclo[2.2.1]heptane-1-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Carboxybicyclo[2.2.1]heptane, Norbornane-1-carboxylic acid
OR22825-100MG
OR22825-250MG
OR22825-500MG
APOSOR22825-100MGEA
237
EUR
APOSOR22825-100MG
APOSOR22825-250MG
APOSOR22825-500MG
Bicyclo[2.2.1]heptane-1-carboxylic acid
Bicyclo[2.2.1]heptane-1-carboxylic acid
Formel:
C₈H₁₂O₂ Molecular Weight: 140.18 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00180984 CAS-Nummer: 18720-30-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...