2-Aminofuran, N-BOC protected 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-[(tert-Butoxycarbonyl)amino]furan, 2-(Boc-amino)furan, tert-Butyl (fur-2-yl)carbamate
OR22968-1G
OR22968-250MG
APOSOR22968-1GEA
60.9
EUR
APOSOR22968-1G
APOSOR22968-250MG
2-Aminofuran, N-BOC protected 97%
2-Aminofuran, N-BOC protected
Formel:
C₉H₁₃NO₃ Molecular Weight: 183,204 g/mol Schmelzpunkt: 98…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02682023 CAS-Nummer: 56267-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...