3,5-Dimethylisoxazole-4-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-3,5-dimethylisoxazole, 4-Cyano-3,5-dimethyl-1,2-oxazole

OR22975-250MG OR22975-1G OR22975-5G
APOSOR22975-250MGEA 65.4 EUR
APOSOR22975-250MG APOSOR22975-1G APOSOR22975-5G
3,5-Dimethylisoxazole-4-carbonitrile
3,5-Dimethylisoxazole-4-carbonitrile
Formel: C₆H₆N₂O
Molecular Weight: 122,125 g/mol
Schmelzpunkt: 43…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681974
CAS-Nummer: 31301-46-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR