5-Methylisoxazole-4-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Methyl-1,2-oxazole-4-sulphonyl chloride, 4-(Chlorosulphonyl)-5-methylisoxazole, 5-Methyl-4-isoxazolesulphonyl chloride

OR22976-250MG
APOSOR22976-250MGEA 21.9 EUR
APOSOR22976-250MG
5-Methylisoxazole-4-sulphonyl chloride
5-Methylisoxazole-4-sulphonyl chloride
Formel: C₄H₄ClNO₃S
Molecular Weight: 181.597 g/mol
Siedepunkt: 56…58 °C (0.7 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02681975
CAS-Nummer: 321309-26-6
UN: 3265
ADR: 8 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR