(5-Methyl-3-phenylisoxazol-4-yl)methylamine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(5-Methyl-3-phenyl-1,2-oxazol-4-yl)methylamine, 4-(Aminomethyl)-5-methyl-3-phenylisoxazole
OR22986-1G
OR22986-5G
OR22986-250MG
APOSOR22986-1GEA
165
EUR
APOSOR22986-1G
APOSOR22986-5G
APOSOR22986-250MG
(5-Methyl-3-phenylisoxazol-4-yl)methylamine 97%
(5-Methyl-3-phenylisoxazol-4-yl)methylamine
Formel:
C₁₁H₁₂N₂O Molecular Weight: 188,226 g/mol Schmelzpunkt: 66…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677687 CAS-Nummer: 306935-01-3 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...