3-(Aminomethyl)-2,5-dimethylfuran 90%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2,5-Dimethylfur-3-yl)methylamine
OR22999-1G
OR22999-250MG
APOSOR22999-1GEA
429
EUR
APOSOR22999-1G
APOSOR22999-250MG
3-(Aminomethyl)-2,5-dimethylfuran 90%
3-(Aminomethyl)-2,5-dimethylfuran
Formel:
C₇H₁₁NO Molecular Weight: 125,168 g/mol Siedepunkt: 42…56 °C (0,1 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02677701 CAS-Nummer: 306934-85-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...