5-Bromo-1-(phenylsulphonyl)-1H-indole 95%

Lieferant: Apollo Scientific
Achtung

OR23041-1G OR23041-5G OR23041-25G
APOSOR23041-1GEA 75.4 EUR
APOSOR23041-1G APOSOR23041-5G APOSOR23041-25G
5-Bromo-1-(phenylsulphonyl)-1H-indole 95%
5-Bromo-1-(phenylsulphonyl)-1H-indole
Formel: C₁₄H₁₀BrNO₂S
Molecular Weight: 336.204 g/mol
Schmelzpunkt: 90…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086087
CAS-Nummer: 118757-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR