1-(Bromomethyl)isoquinoline hydrobromide

Lieferant: Apollo Scientific
Gefahr

OR23078-1G OR23078-250MG OR23078-5G
APOSOR23078-1GEA 142 EUR
APOSOR23078-1G APOSOR23078-250MG APOSOR23078-5G
1-(Bromomethyl)isoquinoline hydrobromide
1-(Bromomethyl)isoquinoline hydrobromide
Formel: C₁₀H₈BrN·BrH
Molecular Weight: 302,993 g/mol
Schmelzpunkt: 210 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086096
CAS-Nummer: 337508-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR