(4-Benzylmorpholin-2-yl)methanol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: {[2-(Hydroxymethyl)morpholin-4-yl]methyl}benzene, 4-Benzyl-2-(hydroxymethyl)morpholine

OR23185-1G OR23185-250MG
APOSOR23185-1GEA 72.5 EUR
APOSOR23185-1G APOSOR23185-250MG
(4-Benzylmorpholin-2-yl)methanol 95%
(4-Benzylmorpholin-2-yl)methanol
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207,269 g/mol
Siedepunkt: 114…116 °C (0,05 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD02682009
CAS-Nummer: 40987-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR