1-[(Benzyloxy)carbonyl]indoline-2-carboxylic acid 90%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-1H-indole-1,2-dicarboxylic acid 1-benzyl ester

OR23215-1G OR23215-5G
APOSOR23215-1GEA 72.5 EUR
APOSOR23215-1G APOSOR23215-5G
1-[(Benzyloxy)carbonyl]indoline-2-carboxylic acid 90%
1-[(Benzyloxy)carbonyl]indoline-2-carboxylic acid
Formel: C₁₇H₁₅NO₄
Molecular Weight: 297,305 g/mol
Schmelzpunkt: 103…105 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682032
CAS-Nummer: 117483-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR