5-Methyl-1-phenyl-1H-pyrazole-4-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(Chlorosulphonyl)-1-phenyl-1H-pyrazole, (4-Chlorosulphonyl-5-methyl-1H-pyrazol-1-yl)benzene

OR23245-1G OR23245-5G
APOSOR23245-1GEA 364 EUR
APOSOR23245-1G APOSOR23245-5G
5-Methyl-1-phenyl-1H-pyrazole-4-sulphonyl chloride
5-Methyl-1-phenyl-1H-pyrazole-4-sulphonyl chloride
Formel: C₁₀H₉ClN₂O₂S
Molecular Weight: 256.709 g/mol
Schmelzpunkt: 57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681919
CAS-Nummer: 342405-38-3
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR