2-Phenyl-1,3-thiazole-4-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR23283-1G OR23283-250MG
APOSOR23283-1GEA 180 EUR
APOSOR23283-1G APOSOR23283-250MG
2-Phenyl-1,3-thiazole-4-carbonyl chloride
2-Phenyl-1,3-thiazole-4-carbonyl chloride
Formel: C₁₀H₆ClNOS
Molecular Weight: 223,679 g/mol
Schmelzpunkt: 100 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02681968
CAS-Nummer: 36094-04-9
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR