6-Phenoxypyridine-3-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-(Chlorosulphonyl)-2-phenoxypyridine

OR23318-250MG
APOSOR23318-250MGEA 0 EUR
APOSOR23318-250MG
6-Phenoxypyridine-3-sulphonyl chloride
6-Phenoxypyridine-3-sulphonyl chloride
Formel: C₁₁H₈ClNO₃S
Molecular Weight: 269,704 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02681949
CAS-Nummer: 368869-91-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR