5-Iodo-2-phenoxypyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR23322-1G
OR23322-10G
APOSOR23322-1GEA
151
EUR
APOSOR23322-1G
APOSOR23322-10G
5-Iodo-2-phenoxypyridine
5-Iodo-2-phenoxypyridine
Formel:
C₁₁H₈INO Molecular Weight: 297,092 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02681952 CAS-Nummer: 352018-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...