3-Methyl-5-(4-methyl-1,2,3-thiadiazol-5-yl)isoxazole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR23346-250MG
OR23346-1G
APOSOR23346-250MGEA
145
EUR
APOSOR23346-250MG
APOSOR23346-1G
3-Methyl-5-(4-methyl-1,2,3-thiadiazol-5-yl)isoxazole-4-carboxylic acid
3-Methyl-5-(4-methyl-1,2,3-thiadiazol-5-yl)isoxazole-4-carboxylic acid
Formel:
C₈H₇N₃O₃S Molecular Weight: 225.224 g/mol Schmelzpunkt: 224…227 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126302 CAS-Nummer: 263385-59-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...