3-(1H-Pyrrol-1-yl)benzaldehyde

Lieferant: Apollo Scientific
Achtung

OR23349-10G OR23349-1G OR23349-250MG
APOSOR23349-10GEA 899 EUR
APOSOR23349-10G APOSOR23349-1G APOSOR23349-250MG
3-(1H-Pyrrol-1-yl)benzaldehyde
3-(1H-Pyrrol-1-yl)benzaldehyde
Formel: C₁₁H₉NO
Molecular Weight: 171,195 g/mol
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086140
CAS-Nummer: 129747-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR