1-[3-(Bromomethyl)phenyl]-1H-pyrrole
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Pyrrol-1-ylbenzyl bromide
OR23350-1G
OR23350-250MG
APOSOR23350-1GEA
215
EUR
APOSOR23350-1G
APOSOR23350-250MG
1-[3-(Bromomethyl)phenyl]-1H-pyrrole
1-[3-(Bromomethyl)phenyl]-1H-pyrrole
Formel:
C₁₁H₁₀BrN Molecular Weight: 236,108 g/mol Schmelzpunkt: 47…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086141 CAS-Nummer: 112596-36-8 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...