2-Chloro-4-hydroxybenzoic acid hydrate 98+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Carboxy-3-chlorophenol hydrate

OR2386-1G OR2386-5G
APOSOR2386-1GEA 58 EUR
APOSOR2386-1G APOSOR2386-5G
2-Chloro-4-hydroxybenzoic acid hydrate 98+%
2-Chloro-4-hydroxybenzoic acid hydrate
Formel: C₇H₅ClO₃·xH₂O
Molecular Weight: 172,57 g/mol (anhy)
Schmelzpunkt: 207…209 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00798116
CAS-Nummer: 440123-65-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR