Methyl 3-hydrazinothiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR24341-10G OR24341-1G
APOSOR24341-10GEA 538 EUR
APOSOR24341-10G APOSOR24341-1G
Methyl 3-hydrazinothiophene-2-carboxylate
Methyl 3-hydrazinothiophene-2-carboxylate
Formel: C₆H₈N₂O₂S
Schmelzpunkt: 69…71 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052080
CAS-Nummer: 75681-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR