Methyl 2-{[3-(chloromethyl)benzoyl]amino}thiophene-3-carboxylate
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-{[3-(Methoxycarbonyl)thiophen-2-yl]carbamoyl}benzyl chloride, Methyl 2-{[3-(chloromethyl)benzoyl]amino}-3-thenoate
OR24396-100MG
OR24396-500MG
APOSOR24396-100MGEA
56.7
EUR
APOSOR24396-100MG
APOSOR24396-500MG
Methyl 2-{[3-(chloromethyl)benzoyl]amino}thiophene-3-carboxylate
Methyl 2-{[3-(chloromethyl)benzoyl]amino}thiophene-3-carboxylate
Formel:
C₁₄H₁₂ClNO₃S Molecular Weight: 309.768 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00728793 CAS-Nummer: 648859-85-2 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...