1-Methylpiperidine-2-carboxylic acid hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Carboxy-1-methylpiperidine hydrochloride

OR24941-1G OR24941-5G
APOSOR24941-1GEA 21.9 EUR
APOSOR24941-1G APOSOR24941-5G
1-Methylpiperidine-2-carboxylic acid hydrochloride 98%
1-Methylpiperidine-2-carboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179.645 g/mol
Schmelzpunkt: 203…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01570459
CAS-Nummer: 25271-35-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR