3-(4-Acetylpiperazin-1-yl)benzoic acid

Lieferant: Apollo Scientific

OR2499-1G OR2499-50MG OR2499-250MG OR2499-500MG
APOSOR2499-1GEA 654 EUR
APOSOR2499-1G APOSOR2499-50MG APOSOR2499-250MG APOSOR2499-500MG
3-(4-Acetylpiperazin-1-yl)benzoic acid
3-(4-Acetylpiperazin-1-yl)benzoic acid
Formel: C₁₃H₁₆N₂O₃
Molecular Weight: 248,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05862615
CAS-Nummer: 896516-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR