2-[(4-Chlorophenyl)sulphonyl]-3-ethoxyacrylonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR25042-1G
OR25042-250MG
OR25042-5G
APOSOR25042-1GEA
106
EUR
APOSOR25042-1G
APOSOR25042-250MG
APOSOR25042-5G
2-[(4-Chlorophenyl)sulphonyl]-3-ethoxyacrylonitrile
2-[(4-Chlorophenyl)sulphonyl]-3-ethoxyacrylonitrile
Formel:
C₁₁H₁₀ClNO₃S Molecular Weight: 271,72 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067998 CAS-Nummer: 32083-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...