S-Methylisothiourea hydroiodide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl carbamimidothioate hydroiodide, Methyl aminomethanimidothioate hydroiodide
OR25052-1G
OR25052-25G
OR25052-5G
APOSOR25052-1GEA
21.9
EUR
APOSOR25052-1G
APOSOR25052-25G
APOSOR25052-5G
S-Methylisothiourea hydroiodide
S-Methylisothiourea hydroiodide
Formel:
C₂H₆N₂S·HI Molecular Weight: 218,06 g/mol Schmelzpunkt: 113…117 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00035598 CAS-Nummer: 4338-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...