3-Amino-4-(phenylsulphonyl)thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR25138-1G OR25138-5G
APOSOR25138-1GEA 154 EUR
APOSOR25138-1G APOSOR25138-5G
3-Amino-4-(phenylsulphonyl)thiophene-2-carboxylic acid
3-Amino-4-(phenylsulphonyl)thiophene-2-carboxylic acid
Formel: C₁₁H₉NO₄S₂
Molecular Weight: 283,323 g/mol
Schmelzpunkt: 163…166 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00067913
CAS-Nummer: 175201-59-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR