5-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1,3,4-oxadiazole-2-thiol

Lieferant: Apollo Scientific

OR25384-1G OR25384-5G
APOSOR25384-1GEA 87 EUR
APOSOR25384-1G APOSOR25384-5G
5-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1,3,4-oxadiazole-2-thiol
5-[(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methyl]-1,3,4-oxadiazole-2-thiol
Formel: C₈H₉BrN₄OS
Molecular Weight: 289,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052137
CAS-Nummer: 175202-84-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR