4-[(4-Chlorophenyl)sulphanyl]thiophene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Carboxythien-3-yl 4-chlorophenyl sulphide, 4-[(4-Chlorophenyl)sulphanyl]-3-thenoic acid
OR25411-1G
OR25411-250MG
OR25411-5G
APOSOR25411-1GEA
201
EUR
APOSOR25411-1G
APOSOR25411-250MG
APOSOR25411-5G
4-[(4-Chlorophenyl)sulphanyl]thiophene-3-carboxylic acid
4-[(4-Chlorophenyl)sulphanyl]thiophene-3-carboxylic acid
Formel:
C₁₁H₇ClO₂S₂ Molecular Weight: 270.755 g/mol Schmelzpunkt: 195…198 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00052149 CAS-Nummer: 149609-86-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...