4,4-Dimethyl-1,3-oxazolidine-2-thione

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4-Dimethyl-2-thioxo-1,3-oxazolidine, 4,4-Dimethyloxazolidine-2-thione

OR25431-1G OR25431-5G OR25431-25G
APOSOR25431-1GEA 126 EUR
APOSOR25431-1G APOSOR25431-5G APOSOR25431-25G
4,4-Dimethyl-1,3-oxazolidine-2-thione
4,4-Dimethyl-1,3-oxazolidine-2-thione
Formel: C₅H₉NOS
Molecular Weight: 131,196 g/mol
Schmelzpunkt: 124…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052156
CAS-Nummer: 54013-55-7
EINECS: 258-924-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR