4-[(2-Methylbut-3-yn-2-yl)oxy]benzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR25514-1G
OR25514-250MG
APOSOR25514-1GEA
232
EUR
APOSOR25514-1G
APOSOR25514-250MG
4-[(2-Methylbut-3-yn-2-yl)oxy]benzonitrile
4-[(2-Methylbut-3-yn-2-yl)oxy]benzonitrile
Formel:
C₁₂H₁₁NO Schmelzpunkt: 30…32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174270 CAS-Nummer: 33143-92-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...