3-[(1-Benzylpiperidin-4-yl)oxy]propanamide

Lieferant: Apollo Scientific
Achtung

OR25625-1G OR25625-5G
APOSOR25625-1GEA 0 EUR
APOSOR25625-1G APOSOR25625-5G
3-[(1-Benzylpiperidin-4-yl)oxy]propanamide
3-[(1-Benzylpiperidin-4-yl)oxy]propanamide
Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262,347 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00178753
CAS-Nummer: 175203-67-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR