2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)pyridine-3-carboxamide, 2-Chloro-N-(2-{[(fur-2-yl)methyl]sulphanyl}ethyl)pyridine-3-carboxamide
OR25742-250MG
OR25742-50MG
APOSOR25742-250MGEA
116
EUR
APOSOR25742-250MG
APOSOR25742-50MG
2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide
2-Chloro-N-(2-{[(fur-2-yl)methyl]thio}ethyl)nicotinamide
Formel:
C₁₃H₁₃ClN₂O₂S Molecular Weight: 296.773 g/mol Schmelzpunkt: 42…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01566566 CAS-Nummer: 680211-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...