(6-Methoxybenzo[b]furan-3-yl)acetic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(6-Methoxybenzofuran-3-yl)acetic acid, 3-(Carboxymethyl)-6-methoxybenzo[b]furan

OR25758-10G OR25758-1G
APOSOR25758-10GEA 403 EUR
APOSOR25758-10G APOSOR25758-1G
(6-Methoxybenzo[b]furan-3-yl)acetic acid 97%
(6-Methoxybenzo[b]furan-3-yl)acetic acid
Formel: C₁₁H₁₀O₄
Molecular Weight: 206,195 g/mol
Schmelzpunkt: 124…127 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677734
CAS-Nummer: 69716-05-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR