N-Benzyl-2-nitro-3,4,5,6-tetrachloroaniline
Lieferant: Apollo Scientific
|
Achtung
|
OR25837-1G
OR25837-5G
APOSOR25837-1GEA
139
EUR
APOSOR25837-1G
APOSOR25837-5G
N-Benzyl-2-nitro-3,4,5,6-tetrachloroaniline
N-Benzyl-2-nitro-3,4,5,6-tetrachloroaniline
Formel:
C₁₃H₈Cl₄N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114878 CAS-Nummer: 69035-11-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...