N1-(4-Chloro-3-nitrophenyl)piperidine-1-carboxamide

Lieferant: Apollo Scientific

OR25866-1G OR25866-5G OR25866-10G
APOSOR25866-1GEA 0 EUR
APOSOR25866-1G APOSOR25866-5G APOSOR25866-10G
N1-(4-Chloro-3-nitrophenyl)piperidine-1-carboxamide
N1-(4-Chloro-3-nitrophenyl)piperidine-1-carboxamide
Formel: C₁₂H₁₄ClN₃O₃
Molecular Weight: 283,711 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00126338
CAS-Nummer: 680212-54-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR