N1-(4-Chloro-3-nitrophenyl)pyrrolidine-1-carboxamide

Lieferant: Apollo Scientific

OR25868-1G OR25868-5G OR25868-10G
APOSOR25868-1GEA 0 EUR
APOSOR25868-1G APOSOR25868-5G APOSOR25868-10G
N1-(4-Chloro-3-nitrophenyl)pyrrolidine-1-carboxamide
N1-(4-Chloro-3-nitrophenyl)pyrrolidine-1-carboxamide
Formel: C₁₁H₁₂ClN₃O₃
Molecular Weight: 269,684 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00126342
CAS-Nummer: 680212-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR