6-Aminobenzo[b]thiophene 1,1-dioxide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Amino-1,1-dioxobenzo[b]thiophene, 6-Amino-1,1-dioxo-1-benzothiophene
OR26007-1G
APOSOR26007-1GEA
121
EUR
APOSOR26007-1G
6-Aminobenzo[b]thiophene 1,1-dioxide
6-Aminobenzo[b]thiophene 1,1-dioxide
Formel:
C₈H₇NO₂S Molecular Weight: 181,212 g/mol Schmelzpunkt: 150…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173800 CAS-Nummer: 20503-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...