1-Amino-2-hydroxyindane

Lieferant: Apollo Scientific

Synonyme: 1-Amino-2-indanol, 1-Aminoindan-2-ol, 2-Hydroxyindan-1-amine, 1-Amino-2,3-dihydro-1H-inden-2-ol

OR2608-1G OR2608-5G OR2608-250MG OR2608-500MG
APOSOR2608-1GEA 157 EUR
APOSOR2608-1G APOSOR2608-5G APOSOR2608-250MG APOSOR2608-500MG
1-Amino-2-hydroxyindane
1-Amino-2-hydroxyindane
Formel: C₉H₁₁NO
Molecular Weight: 149.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00053472
CAS-Nummer: 74165-73-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR