2,6-Diaminopyrimidin-4(3H)-one 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Diamino-1,6-dihydro-6-oxopyrimidine

OR26562-1G OR26562-250MG
APOSOR26562-1GEA 24.8 EUR
APOSOR26562-1G APOSOR26562-250MG
2,6-Diaminopyrimidin-4(3H)-one 98%
2,6-Diaminopyrimidin-4(3H)-one
Formel: C₄H₆N₄O
Molecular Weight: 126,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00043013
CAS-Nummer: 143504-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR