DL-Phenylglycine ethyl ester hydrochloride 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl amino(phenyl)acetate hydrochloride, α-(Ethoxycarbonyl)benzylamine hydrochloride, H-DL-2-Phg-OEt·HCl, Ethyl 2-amino-2-phenylacetate hydrochloride
OR26810-1G
OR26810-5G
OR26810-25G
APOSOR26810-1GEA
102
EUR
APOSOR26810-1G
APOSOR26810-5G
APOSOR26810-25G
DL-Phenylglycine ethyl ester hydrochloride 98%
DL-Phenylglycine ethyl ester hydrochloride
Formel:
C₁₀H₁₃NO₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114583 CAS-Nummer: 879-48-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...