3-Bromo-N-(4-bromophenyl)propanamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-N-(4-bromophenyl)propionamide, 4-Bromo-N-(3-bromopropanoyl)aniline

OR26966-250MG OR26966-500MG
APOSOR26966-250MGEA 198 EUR
APOSOR26966-250MG APOSOR26966-500MG
3-Bromo-N-(4-bromophenyl)propanamide
3-Bromo-N-(4-bromophenyl)propanamide
Formel: C₉H₉Br₂NO
Molecular Weight: 306.982 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00124398
CAS-Nummer: 7661-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR