2-(Morpholin-4-yl)acetamide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Morpholinoacetamide, 2-(4-Morpholinyl)acetamide
OR27100-100MG
OR27100-250MG
APOSOR27100-100MGEA
37.7
EUR
APOSOR27100-100MG
APOSOR27100-250MG
2-(Morpholin-4-yl)acetamide
2-(Morpholin-4-yl)acetamide
Formel:
C₆H₁₂N₂O₂ Molecular Weight: 144,172 g/mol Schmelzpunkt: 122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00465044 CAS-Nummer: 5625-98-9 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...