3,5-Dichloro-4-hydrazinopyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: (3,5-Dichloropyridin-4-yl)hydrazine

OR27149-1G OR27149-500MG OR27149-5G
APOSOR27149-1GEA 284 EUR
APOSOR27149-1G APOSOR27149-500MG APOSOR27149-5G
3,5-Dichloro-4-hydrazinopyridine
3,5-Dichloro-4-hydrazinopyridine
Formel: C₅H₅Cl₂N₃
Molecular Weight: 178,019 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00174279
CAS-Nummer: 153708-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR