4-Cyclopropyl-6-(methylthio)-1,3,5-triazin-2-amine

Lieferant: Apollo Scientific
Achtung

OR27308-1G OR27308-5G
APOSOR27308-1GEA 109 EUR
APOSOR27308-1G APOSOR27308-5G
4-Cyclopropyl-6-(methylthio)-1,3,5-triazin-2-amine
4-Cyclopropyl-6-(methylthio)-1,3,5-triazin-2-amine
Formel: C₇H₁₀N₄S
Molecular Weight: 182,246 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052768
CAS-Nummer: 175204-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR